ContactUsLinkComponent
ImmPrint™ Permanent Marking Pen, Vector Laboratories
ImmPrint™ Permanent Marking Pen, Vector Laboratories
Catalog # 101098-070
undefined
ImmPrint™ Permanent Marking Pen, Vector Laboratories
Catalog # 101098-070
Supplier Number:  H-6100
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Permanent Histology Marking Pen, set of 5
  • Cat. No.:
    101098-070
  • Supplier no.:
    H-6100
Specifications
About this item
An all-purpose marking pen that's designed for writing on slides, with marks that remain legible through histology processing.
  • Solvent-resistant permanent marking pen
  • Designed for writing on frosted glass microscope slides, tissue cassettes, and most hard surfaces
  • High density, fast-drying, black ink is resistant to most organic solvents
  • Has a smooth writing tip that resists drying out
The ImmPrint™ Histology Pen is a solvent-resistant permanent marking pen designed for writing on frosted glass microscope slides, tissue cassettes, and most hard surfaces. The high density, fast-drying, black ink is resistant to most organic solvents encountered in histological applications such as alcohols and clearing agents. Unlike other pens commonly used for histology, the ImmPrint™ pen has a smooth writing tip that resists drying out.