ContactUsLinkComponent
DBI-SALA® Rope Lifeline with Snap Hooks, ORS Nasco
DBI-SALA® Rope Lifeline with Snap Hooks, ORS Nasco
  76111-338
DBI-SALA® Rope Lifeline with Snap Hooks, ORS Nasco
  76111-338
 :  098-1202794
  • Length:
    50'
  • Material:
    Polyester/Polypropylene Blend
  • Cat. No.:
    76111-338
  • Load capacity:
    310 lbs.
Ideal for use as a vertical lifeline system with a rope grab.
  • Equipped with i-Safe™ RFID identification tag for recording and storing information on inspections
  • Durable, high strength 5-tuck splice provides an extra layer of ruggedness, strength, and field longevity
  • Polyester/polypropylene blend offers additional abrasion and UV resistance for added durability and longevity
Built-in self locking snap hook at one end offers fast and safe connection to an anchorage connector, such as a tie-off adaptor for added productivity.
 : OSHA 1910.66, OSHA 1926.502, Capital Safety General Manufacturing Requirements, ANSI A10.32, ANSI Z359.1.