DBI-SALA® Nano-Lok™ Twin Leg Tie-Back SLR with Twin Quick Connector, ORS Nasco
Supplier: Safety & Industrial Supplies
Certificates
About this item
Ergonomically designed for ease-of-use, this unit attaches directly to back of most harnesses.
- Tie-back leg eliminates the need for an anchor connector, tie-off adapter, or scaffold choker
- Compact, lightweight design
- Design fits snugly against worker’s back, eliminating nuisance “flopping"
- High-strength housing provides maximum durability
- Automatic quick-activating arrestor stops falling worker within inches and reduces average arresting forces to 900 lbs. or less
- Equipped with i-Safe™
- Installs to web, leaves dorsal D-ring open for other equipment or rescue
Certifications: ANSI Z359.14, OSHA 1910.66, OSHA 1926.502, Capital Safety Gen. Mfg. Req., ANSI A10.3.
Frequently Bought Together