ContactUsLinkComponent
DBI-SALA® ExoFit™ Vest-Style Positioning Harness with Back and Front D-Rings, ORS Nasco
DBI-SALA® ExoFit™ Vest-Style Positioning Harness with Back and Front D-Rings, ORS Nasco
Catalog # 76111-156
Supplier:  Avantor
DBI-SALA® ExoFit™ Vest-Style Positioning Harness with Back and Front D-Rings, ORS Nasco
Catalog # 76111-156
Supplier:  Avantor
Specifications
  • Color:
    Blue/Gray
  • Description:
    Vest-Style Positioning Harness with Quick Connect Buckles
  • Material:
    Polyester Strap
  • Size:
    XX-Large
  • Cat. No.:
    76111-156
  • Load capacity:
    420 lbs.
  • Connector:
    Back and side D-rings
Specifications
About this item
Lightweight, moisture wicking, breathable padding with soft edging wraps around the wearer for the ultimate in comfort, security, and no-tangle donning.
  • Back D-ring provides a safe connection for a fall arrest system, and keeps users upright every time
  • Hip D-rings are ideal for positioning, to hold and sustain users at a specific work location
  • Soft, lightweight webbing is extremely strong, yet isn't stiff or coarse for added comfort and durability
  • Built-in reflective piping enhances visibility in low light conditions
  • Parachute torso buckles complete the 5-point adjustment system for a perfect fit
Anti-tear ballistic nylon is abrasion-resistant for rugged strength and durability, and has twice the endurance of standard nylon or polyester. A Velcro® cover protects harness labels and holds an i-Safe™ identification tag for recording and storing information on inspections.