Acriflavin-Cefsulodin-Vancomycin (ACV) Supplement, Himedia®
Catalog # 10662-034
Supplier: HiMedia
CAS Number:
Specifications
- Description:ACV Supplement
- Size:5 vials
- Cat. No.:10662-034
- Supplier no.:FD284-5VL
Specifications
About this item
ACV supplement is used for the isolation of Enterohemorrhagic E
One vial is sufficient for 500 mL medium.
coli (EHEC).
Caution: Store at 2 to 8 °C. For medicinal use only.