GloMax Explorer Absorbance Module Upgrade, Promega
Catalog # PAGM3520
Supplier: Promega Corporation
Specifications
- Description:GloMax Explorer Absorbance Module Upgrade, 1 each
- Size:1 each
- Cat. No.:PAGM3520
- Storage temperature:none
- Supplier no.:GM3520
Specifications
About this item
These accessories are intended for use with the GloMax Discover and Explorer Systems.
- For use with GloMax(R) Discover and Explorer Systems
- Filter Paddles only available for GloMax(R) Discover System
- Absorbance Module only available for GloMax(R) Explorer System
These accessories are intended for use with the GloMax Discover and Explorer Systems.