MagneSphere Technology Magnetic Separation Stand (two-position), Promega
Supplier: Promega Corporation
Certificates
About this item
Use in conjunction with the PolyATtract Systems. Ideal for applications requiring multiple paramagnetic isolations of biomolecules.
- Use with both MagneSphere(R) and PolyATtract(R) System components
- Conduct simultaneous processing of the contents of two tubes or twelve tubes of various sizes
The MagneSphere Technology Magnetic Separation Stands can be used in conjunction with any of the PolyATtract Systems and are ideal for applications requiring multiple paramagnetic isolations of biomolecules. These stands use the same strong rare earth magnet used in the PolyATtract Systems Magnetic Separation Stands and come in a variety of sizes to accommodate 2-96 samples.
Specifications
- Catalog No:
- PAZ5331
- PAZ5332
- PAZ5333
- Storage Temperature:
- 15°C to 30°C
- 15°C to 30°C
- 15°C to 30°C