Order Entry
United States
ContactUsLinkComponent
pGL4.32[luc2P/NF-kappa-B-RE/Hygro] Vector, 20 µg, Promega
pGL4.32[luc2P/NF-kappa-B-RE/Hygro] Vector, 20 µg, Promega
Catalog # PAE8491
CAS Number:  
undefined
pGL4.32[luc2P/NF-kappa-B-RE/Hygro] Vector, 20 µg, Promega
Catalog # PAE8491
Supplier Number:  E8491
CAS Number:  

Specifications

  • Description:
    pGL4.32[luc2P/NF-κB-RE/Hygro] Vector
  • Size:
    20 μg
  • Storage Conditions:
    Store at –20°C unopened. See product insert for individual component storage conditions before and after opening.
  • Cat. No.:
    PAE8491
  • Storage temperature:
    –30...–10 °C
  • Supplier no.:
    E8491

Specifications

About this item

pGL4.44[luc2P/AP1 RE/Hygro], pGL4.49[luc2P/TCF-LEF RE/Hygro], pGL4.29[luc2P/CRE/Hygro], pGL4.30[luc2P/NFAT-RE/Hygro], pGL4.32[luc2P/NF-κB-RE/Hygro], pGL4.33[luc2P/SRE/Hygro] and pGL4.34[luc2P/SRF-RE/Hygro] Vectors can explore various signaling pathways.

  • Reporter Vectors Ready to Use in Your Signaling Pathway Without Cloning
  • Offer a number of response elements that are involved in nuclear response, kinase and other signaling pathways
  • Each vector included an optimized firefly luciferase gene that codes a reporter with a one-hour half-life
  • Hygromycin resistance selectable marker means choosing transient transfection or generating a stable cell line

Creating a cell line with an indicator of a functional signaling pathway is useful for deciphering the components in a signaling pathway. These tools are made by inserting multiple repeats of a response element upstream of a minimal promoter (minP). We offer vectors that report the activity of a variety of cellular stress pathways using the optimized firefly luciferase gene luc2 in the pGL4 backbone. The luc2 gene is engineered to remove most cryptic transcription factor binding sites and improve mammalian expression through codon optimization. The luc2P and luc2CP and RapidResponse genes are luc2 genes appended with degradation sequences to influence the cellular half-life of the luc2 gene. The RapidResponse genes respond more rapidly to stimuli but at the expense of signal intensity. The luc2P (1-hour half-life) gene responds more rapidly than luc2 (3-hour half-life) with moderate signal intensity. These response element vectors also have a hygromycin resistance selectable marker for use either in transient transfection experiments or for selection of a stable cell line. To speed research, several pre-designed response element vectors based on the pGL4.27 Vector are available. Some of these also are available stable cell lines (GloResponse Cell Lines). These available signaling pathway vectors include MAPK/JNK pathway with the pGL4.44[luc2P/AP1 RE/Hygro] Vector, Wnt pathway with the pGL4.49[luc2P/TCF-LEF RE/Hygro] Vector, cAMP/PKA pathway with the pGL4.29[luc2P/CRE/Hygro] Vector, Calcium/Calcineurin pathway with the pGL4.30[luc2P/NFAT-RE/Hygro] Vector, NF-kappaB pathway with the pGL4.32[luc2P/NF-κB-RE/Hygro] Vector, MAP/ERK pathway with the pGL4.33[luc2P/SRE/Hygro] Vector and RhoA pathway with the pGL4.34[luc2P/SRF-RE/Hygro] Vector.