Order Entry
United States
ContactUsLinkComponent
pGEM-5Zf(+) Vector, 20 µg, Promega
pGEM-5Zf(+) Vector, 20 µg, Promega
Catalog # PAP2241
CAS Number:  
undefined
pGEM-5Zf(+) Vector, 20 µg, Promega
Catalog # PAP2241
Supplier Number:  P2241
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    20 μg
  • Cat. No.:
    PAP2241
  • Storage temperature:
    Less than –65 °C
  • Supplier no.:
    P2241

Specifications

About this item

Used as a standard cloning vector and as a template for in vitro transcription.

  • Standard Cloning Vector and Template for In vitro Transcription
  • Blue/white screening to easily identify recombinant clones
  • Multiple cloning site provides a convenient selection of restriction sites for cloning

The pGEM-5Zf(+) Vector serves as a standard cloning vector and as a template for in vitro transcription. This vector contains T7 and SP6 RNA polymerase promoters flanking a multiple cloning region within the alpha-peptide coding region of beta-galactosidase. Insertional inactivation of the alpha-peptide allows recombinant clones to be identified directly by color screening on indicator plates when using appropriate E. coli strains (e.g., JM109). The multiple cloning region contains unique restriction sites for ApaI, AatII, SphI, NcoI, SacII, EcoRV, SpeI, NotI, PstI, SalI, NdeI, SacI, BstXI and NsiI.