ContactUsLinkComponent
Human CD102 / ICAM-2 ELISA Assay Kit, Eagle Biosciences
Human CD102 / ICAM-2 ELISA Assay Kit, Eagle Biosciences
Catalog # 102994-024
Supplier:  Eagle Biosciences
CAS Number:  
Human CD102 / ICAM-2 ELISA Assay Kit, Eagle Biosciences
Catalog # 102994-024
Supplier:  Eagle Biosciences
Supplier Number:  C0231-K01
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Primary antibody reactivity:
    Human
  • Description:
    Human CD102 / ICAM-2 ELISA Assay Kit
  • Size:
    1 kit
  • Cat. No.:
    102994-024
  • Supplier no.:
    C0231-K01
Specifications
About this item
Human sICAM-2 ELISA (CD102) detects sICAM-2 in various samples.
  • Dynamic Range: 0-17 ug/L
  • Sample Size: 180 uL
  • Incubation: 4.75 hours
The Human sICAM-2 ELISA (CD102) is to be used for the in-vitro quantitative determination of soluble Inter Cellular Adhesion Molecule-2 (sICAM-2) in human serum, plasma, buffered solutions or cell culture medium. The assay will recognize both natural and recombinant human sICAM-2. The Eagle Biosciences Human sICAM-2 ELISA/CD102 ELISA Assay kit is for Research Use Only.