Order Entry
United States
ContactUsLinkComponent
IgM (rabbit) ELISA, Eagle Biosciences
IgM (rabbit) ELISA, Eagle Biosciences
Catalog # 102993-478
Supplier:  Eagle Biosciences
CAS Number:  
IgM (rabbit) ELISA, Eagle Biosciences
Catalog # 102993-478
Supplier:  Eagle Biosciences
Supplier Number:  IGM79-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Primary antibody reactivity:
    Rabbit
  • Target protein:
    IgM
  • Description:
    IgM (rabbit) ELISA
  • Size:
    1 kit
  • Cat. No.:
    102993-478
  • Supplier no.:
    IGM79-K01

Specifications

About this item

Rabbit IgM ELISA is designed to detect rabbit IgM in complex samples.

  • Dynamic Range: 4.69 - 300 ng/ml
  • Sensitivity: 4.69 ng/ml
  • Incubation: 2.5 house

The Eagle Biosciences Rabbit IgM ELISA Assay Kit is designed for the quantitative determination of rabbit IgM in complex samples (serum or other biological samples). The Rabbit IgM ELISA Assay Kit is for research use only and should not be used in diagnostic procedures.