Order Entry
United States
ContactUsLinkComponent
FGF23 (C-terminal) ELISA Assay Kit, Eagle Biosciences, Inc.
FGF23 (C-terminal) ELISA Assay Kit, Eagle Biosciences, Inc.
Catalog # 102993-984
Supplier:  Eagle Biosciences
CAS Number:  
FGF23 (C-terminal) ELISA Assay Kit, Eagle Biosciences, Inc.
Catalog # 102993-984
Supplier:  Eagle Biosciences
Supplier Number:  BI-20702
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    FGF23 (C-terminal) ELISA Assay Kit
  • Size:
    1 kit
  • Cat. No.:
    102993-984
  • Supplier no.:
    BI-20702

Specifications

About this item

FGF23 (Fibroblast Growth Factor 23) ELISA detects FGF23 in serum or plasma.

  • Dynamic Range: 0.63 - 10 nmol/L
  • Sensitivity: 0.05 nmol/L
  • Incubation: 3.5 hours

FGF23 (fibroblast growth factor 23) is a 32 kDa protein with 251 amino acids that is proteolytically processed between arginine179 and serine180 to generate N-terminal and C-terminal fragments. FGF23 is mainly secreted by osteocytes and controls phosphate and 1,25(OH)2 vitamin D homeostasis.