Order Entry
United States
ContactUsLinkComponent
15-Epi-Lipoxin A4 ELISA Kit, Eagle Biosciences, Inc.
15-Epi-Lipoxin A4 ELISA Kit, Eagle Biosciences, Inc.
Catalog # 102993-080
Supplier:  Eagle Biosciences
CAS Number:  
15-Epi-Lipoxin A4 ELISA Kit, Eagle Biosciences, Inc.
Catalog # 102993-080
Supplier:  Eagle Biosciences
Supplier Number:  15E39-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Target protein:
    15-Epi-Lipoxin A4
  • Description:
    15-Epi-Lipoxin A4 ELISA Kit
  • Size:
    1 kit
  • Cat. No.:
    102993-080
  • Supplier no.:
    15E39-K01

Specifications

About this item

15-Epi-Lipoxin A4 ELISA is to detect of 15-Epi-Lipoxin A4 in biological fluids.

  • Dynamic Range: Cut-Off
  • Sensitivity: Cut-off
  • Incubation: 2 hours

15-epi-Lipoxin A4 (15-epi-LXA4) is an aspirin-triggered eicosanoid believed to be involved in the positive attributes of aspirin therapy for heart, cancer, and human immunodeficiency virus patients. During inflammation, neutrophils are activated. 15-epi-LXA4, when administered in vivo, inhibits neutrophil activation and dampens inflammation (Takano, 1997). 15-epi-LXA4 is naturally formed in the body. The development of this assay, which is both sensitive and selective for 15-epi-LXA4, will be essential to researchers studying anti-inflammatory medications.