- Description:Dopamine ELISA
- Size:1 kit
- Cat. No.:102994-258
- Supplier no.:DOP31-K01
Dopamine ELISA detects Dopamine in urine or plasma by a microplate enzyme immuno .
- Dynamic Range: 1.5 - 2560 ng/ml
- Sensitivity: Urine: 0.46 ng/ml, Plasma: 23 pg/ml
- Incubation: Overnight
Catecholamine is the name of a group of aromatic amines (noradrenaline, adrenaline, dopamine, and their derivatives) which act as hormones and neurotransmitter, respectively. Adrenaline and noradrenaline are formed from dopamine. They act on the cardiac musculature and the metabolism (adrenaline) as well as on the peripheral circulation (noradrenaline) and help the body to cope with acute and chronic stress.