- Description:Histamine ELISA
- Size:1 kit
- Cat. No.:102994-462
- Supplier no.:HIS31-K01
Histamine ELISA is intended to detect histamine in Plasma, Urine, and Cell Culture.
- Dynamic Range: 0.15 - 25 ng/mL
- Sensitivity: 0.02 ng/mL
- Incubation: 2.5 hours
Histamine plays a major role in the initial phase of an anaphylactic reaction. The quantification of histamine in plasma after allergen administration is of clinical interest. Histamine is part of the immune response to foreign pathogens and it increases the permeability of the capillaries to white blood cells and other proteins in order to allow them to engage foreign invaders in the affected tissues. Responsible for the biological effects of histamine in tissue are the activation of different surface receptors, for instance H1, H2 and H3. Histamine is involved in the regulating physiological function in the gut and acting as a neurotransmitter.