Order Entry
United States
ContactUsLinkComponent
 
OxyStat ELISA, Eagle Biosciences
Catalog # 102993-996
Supplier:  Eagle Biosciences
CAS Number:  
OxyStat ELISA, Eagle Biosciences
Catalog # 102993-996
Supplier:  Eagle Biosciences
Supplier Number:  BI-5007
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    OxyStat ELISA
  • Size:
    1 kit
  • Cat. No.:
    102993-996
  • Supplier no.:
    BI-5007

Specifications

About this item

OxyStat Colorimetric detects peroxides in plasma, serum and cell culture supernatants.

  • Dynamic Range: 7 - 600 µmol/l
  • Sensitivity: 7 µmol/l
  • Incubation: 15 minutes

Cells and tissues are sensitive to oxidative stress, caused by the formation of free radicals. If not deactivated by antioxidants, organic peroxides and hydroperoxides are the first reaction products between cellular constituents and free radicals or other reactive oxygen derivates.