To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Host:
- Primary antibody reactivity:
- Description:Normetanephrine & Metanephrine (Plasma) ELISA Kit
- Size:1 kit
- Environmentally Preferable:
- Cat. No.:102994-810
- Supplier no.:NMN31-K02
Specifications
About this item
Normetanephrine and Metanephrine ELISA detects normetanephrine and metanephrine in plasma.
- Dynamic Range: Metanephrine: 20 - 3000 pg/ml, Normetanephrine: 20 - 3000 pg/ml
- Sensitivity: Metanephrine: 10 pg/ml, Normetanephrine: 15 pg/ml
- Incubation: Overnight
Normetanephrine and metanephrine are physiologically formed from the catechol-amines noradrenaline and adrenaline by the enzyme catechol-O-methyltransferase (COMT). Increased levels of normetanephrine and metanephrine can be found in pheochromocytoma, ganglioneuroma and other neurogenic tumors. The Normetanephrine & Metanephrine (Plasma) ELISA assay kit provides materials for the quantitative measurement of free normetanephrine and metanephrine in EDTA plasma. Normetanephrine and metanephrine are quantitatively acylated to their N-acyl-derivates.