Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Primary antibody reactivity:Pig
- Target protein:IgA
- Description:IgA (Porcine) ELISA
- Size:1 kit
- Cat. No.:102993-440
- Supplier no.:IGA59-K01
Porcine IgA ELISA is designed to detect IgA antibodies in porcine serum and biological samples.
- Dynamic Range: 15.6 ng/ml and 1000 ng/ml
- Sensitivity: 156 ng/ml
- Incubation: 4.5 hours
The Eagle Biosciences Porcine (Pig) IgA ELISA Assay Kit is designed for the quantitative determination of IgA antibodies in porcine serum and biological samples. The Porcine (Pig) IgA ELISA Assay Kit is for research use only and should not be used in diagnostic procedures.