Order Entry
United States
ContactUsLinkComponent
BNP Fragment ELISA, Eagle Biosciences
BNP Fragment ELISA, Eagle Biosciences
Catalog # 102993-988
Supplier:  Eagle Biosciences
CAS Number:  
BNP Fragment ELISA, Eagle Biosciences
Catalog # 102993-988
Supplier:  Eagle Biosciences
Supplier Number:  BI-20852W
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    BNP Fragment ELISA
  • Size:
    1 kit
  • Cat. No.:
    102993-988
  • Supplier no.:
    BI-20852W

Specifications

About this item

BNP Fragment (Nt-proBNP 8-29) ELISA detects human BNP Fragment concentrations in serum and plasma/

  • Dynamic Range: 200 - 6400 pmol/L
  • Sensitivity: 171 pmol/L
  • Incubation: overnight

The natriuretic peptides are members of a family of structurally similar but genetically distinct peptide hormones, consisting of atrial-, brain-, and C-type (ANP, BNP, and CNP, respectively). ANP and BNP preferentially bind to a membrane-bound guanylyl cyclase (GC) receptor called GC-A or NPR1, whereas CNP is the physiological ligand for GC-B (NPR2). The natriuretic peptides play an important role in the regulation of cardiovascular and renal homeostasis and in the regulation of fatty acid metabolism and body weight.