Order Entry
United States
ContactUsLinkComponent
CoproELISA C. difficile GDH, Eagle Biosciences, Inc.
CoproELISA C. difficile GDH, Eagle Biosciences, Inc.
Catalog # 103389-354
Supplier:  Eagle Biosciences
CAS Number:  
CoproELISA C. difficile GDH, Eagle Biosciences, Inc.
Catalog # 103389-354
Supplier:  Eagle Biosciences
Supplier Number:  784-01
CAS Number:  

Specifications

  • Description:
    CoproELISA C. difficile GDH
  • Size:
    1 kit
  • Cat. No.:
    103389-354

Specifications

About this item

CoproELISA C.difficile GDH ELISA detects it in specimens from patients suspected of having C

  • Dynamic Range: Cut-Off
  • Sensitivity: Cut-Off
  • Incubation: 2 hours

The gram-positive anaerobic bacillus Clostridium difficile is the leading causative agent of antibiotic-associated diarrhea and pseudomembranous colitiss. This pathogen is capable of causing disease that could be severe or fatal if not diagnosed on time and treated. Exposure to antibiotics is the major risk factor for C. difficile infection. Infection can develop if the normal gastrointestinal flora is disrupted by antibiotic therapy and a person acquires toxin-producing C. difficile, typically via the fecal-oral route. C. difficile’s key virulence factors are toxin A and toxin B. These toxins show high sequence and functional homology. Toxin A has been described as a tissue damaging enterotoxin which attracts neutrophils and monocytes and toxin B as a potent cytotoxin that degrades the colonic epithelial cells. Most virulent strains produce both toxins, however, toxin A negative/toxin B positive strains are also capable of causing disease. All strains of C. difficile produce high levels of GDH. Therefore, C. difficile’s GDH enzyme is considered a very good antigen marker for detection of this organism. The CoproELISA C. difficile GDH test is a highly specific and sensitive diagnostic kit for GDH in stool specimens. A positive result confirms the presence of C. difficile and a negative result indicates its absence. A separate test should be performed to confirm the presence of C. difficile toxins in the GDH positive samples.

diff.