Order Entry
United States
ContactUsLinkComponent
Swine IL-2 ELISA KIT, Eagle Biosciences
Swine IL-2 ELISA KIT, Eagle Biosciences
Catalog # 102993-492
Supplier:  Eagle Biosciences
CAS Number:  
Swine IL-2 ELISA KIT, Eagle Biosciences
Catalog # 102993-492
Supplier:  Eagle Biosciences
Supplier Number:  IL251-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Target protein:
    IL-2
  • Description:
    Swine IL-2 ELISA KIT
  • Size:
    1 kit
  • Cat. No.:
    102993-492
  • Supplier no.:
    IL251-K01

Specifications

About this item

Swine IL-2 ELISA detects Swine IL-2 concentrations in cell culture supernates, serum, and plasma.

  • Dynamic Range: 3.9 - 125 pg/ml
  • Sensitivity: 2 pg/ml
  • Incubation: 3.0 hours

Interleukin 2 (IL-2) is a pleiotropic cytokine produced primarily by mitogen- or Antigens-activated T lymphocytes (1, 2). Human IL-2 (also known as T-cell growth factor) is produced by T-cells in response to Antigensic or mitogenic stimulation. IL-2 is a potent lymphoid cell growth factor which exerts its biological activity primarily on T cells promoting proliferation and maturation.
IL-2 has been found to stimulate growth and differentiation of B cells, NK cells, LAK cells, monocytes, and oligodendocytes. IL-2 is involved in treatment of cancers such as melanoma and renal cell cancer. It plays a key role in promoting the clonal expansion of Antigens-specific T cells. In addition, IL-2 has also been shown to mediate multiple immune responses on a variety of cell types. At the amino acid sequence level, there is approximately 72% similarity between mature porcine and human IL-2. The biological effects of IL-2 are mediated by specific cell surface receptor complexes. The functional high-affinity receptor for IL-2 is composed of three distinct polypeptide chains (3, 4). IL-2 stimulates the proliferation of thymocytes; stimulates the proliferation and differentiation of activated B cells; promotes the growth, differentiation and cytocidal activity of monocytes; induces the growth of natural killer cells and stimulates cytokine production by these cells as well as the cytolytic activity of these cells; enhances the production of lymphocyte-activated killer (LAK) cells; and induces the proliferation and differentiation of oligodendrocytes (1, 2).