Order Entry
United States
ContactUsLinkComponent
IgY (chicken) ELISA, Eagle Biosciences
IgY (chicken) ELISA, Eagle Biosciences
Catalog # 102993-468
Supplier:  Eagle Biosciences
CAS Number:  
IgY (chicken) ELISA, Eagle Biosciences
Catalog # 102993-468
Supplier:  Eagle Biosciences
Supplier Number:  IGG99-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    IgY (chicken) ELISA
  • Size:
    1 kit
  • Cat. No.:
    102993-468
  • Supplier no.:
    IGG99-K01

Specifications

About this item

IgY (chicken) ELISA detects chicken IgY in complex samples.

  • Dynamic Range: 3.125 - 200 ng/ml
  • Sensitivity: 80 ng/ml (sample dilution 1:25)
  • Incubation: 2.5 hours

The Eagle Biosciences Chicken IgY ELISA Assay Kit is designed for the quantitative determination of chicken IgY in complex samples (serum or other biological samples). The Chicken IgY ELISA Assay Kit is for research use only and should not be used in diagnostic procedures.