Order Entry
United States
ContactUsLinkComponent
Mouse / Rat Active GLP-1 ELISA , Eagle Biosciences
Mouse / Rat Active GLP-1 ELISA , Eagle Biosciences
Catalog # 102994-422
Supplier:  Eagle Biosciences
CAS Number:  
Mouse / Rat Active GLP-1 ELISA , Eagle Biosciences
Catalog # 102994-422
Supplier:  Eagle Biosciences
Supplier Number:  GP121-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugate ELISA:
    HRP
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse / Rat Active GLP-1 ELISA
  • Size:
    1 kit
  • Cat. No.:
    102994-422
  • Supplier no.:
    GP121-K01

Specifications

About this item

Mouse Rat GLP-1 Active (7-36) ELISA detects bioactive GLPe-1 (7-36) amide level in plasma samples.

  • Dynamic Range: .85 - 146 pM
  • Sensitivity: 0.1 pM
  • Incubation: 24.5 hours

The Eagle Biosciences Mouse/Rat Active GLP-1 ELISA Assay Kit (enzyme-linked immunoassay kit) is intended quantitative determination of bioactive glucagon-like peptide-1 (7-36) amide [GLP-1 (7-36)] level in rat and mouse plasma samples with only 20 ul of sample volume. E The primary amino acid sequence of GLP-1 peptide is identical among mammalian species, i.e. rat, mouse, pig, human, etc. Eagle Biosciences Mouse/Rat Active GLP-1 ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.