Order Entry
United States
ContactUsLinkComponent
GLP-1 Total ELISA Assay Kit, Eagle Biosciences, Inc.
GLP-1 Total ELISA Assay Kit, Eagle Biosciences, Inc.
Catalog # 102994-424
Supplier:  Eagle Biosciences
CAS Number:  
GLP-1 Total ELISA Assay Kit, Eagle Biosciences, Inc.
Catalog # 102994-424
Supplier:  Eagle Biosciences
Supplier Number:  GP131-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugate ELISA:
    HRP
  • Description:
    GLP-1 Total ELISA Assay Kit
  • Size:
    1 kit
  • Cat. No.:
    102994-424
  • Supplier no.:
    GP131-K01

Specifications

About this item

GLP-1 Total ELISA determines sum of glucagon-like peptide-1 (7-36) & (9-36) in plasma.

  • Dynamic Range: 2 -54 pmol/L
  • Sensitivity: 0.6 pmol/L
  • Incubation: overnight

The primary amino acid sequence of GLP-1 peptide is identical among mammalian species, i.e. rat, mouse, pig, human, etc. The Eagle Biosciences GLP-1 Total ELISA Assay Kit (enzyme-linked immunoassay kit) is intended quantitative determination of sum value of glucagon-like peptide-1 (7-36) and (9-36) [GLP-1 (7-36)] and (9-36) [GLP-1 (9-36)] in plasma samples. Eagle Biosciences GLP-1 Total ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.