Order Entry
United States
ContactUsLinkComponent
ADIPONECTIN ELISA , Eagle Biosciences, Inc.
ADIPONECTIN ELISA , Eagle Biosciences, Inc.
Catalog # 102994-284
Supplier:  Eagle Biosciences
CAS Number:  
ADIPONECTIN ELISA , Eagle Biosciences, Inc.
Catalog # 102994-284
Supplier:  Eagle Biosciences
Supplier Number:  E09
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Target protein:
    Adiponectin
  • Description:
    ADIPONECTIN ELISA
  • Size:
    1 kit
  • Cat. No.:
    102994-284
  • Supplier no.:
    E09

Specifications

About this item

Adiponectin ELISA measures Adiponectin in the human serum & plasma samples for research use only.

  • Dynamic Range: 2 -100 ng/ml
  • Sensitivity: 0.6 ng/ml
  • Incubation: 1.75 hours

Adiponectin is a 30kDa protein which percentage in serum proteins is 0.01%. It is mainly synthesized by Adipocytes, but also muscle cells and hepatocytes have the ability to synthesize Adiponectin. Until now, IGF-I is the only known natural inductor of the synthesis. It consists of  a Collagen-like N-terminal and a globular C-terminal domain [1]. In vivo Adiponectin appears with different oligomers. Beside the trimer and ditrimer also high molecular multimers exist [1-3]. Up to now two different receptors are known, both receptors are ubiquitary expressed, though the distribution in the tissues varies. The Adiponectin Receptor 1 (AdipoR1) is especially in muscle- and AdipoR2 in liver tissue synthesized [4].