Order Entry
United States
ContactUsLinkComponent
Human CD62E / ELAM-1 / E-Selectin ELISA Assay Kit, Eagle Biosciences
Human CD62E / ELAM-1 / E-Selectin ELISA Assay Kit, Eagle Biosciences
Catalog # 102994-042
Supplier:  Eagle Biosciences
CAS Number:  
Human CD62E / ELAM-1 / E-Selectin ELISA Assay Kit, Eagle Biosciences
Catalog # 102994-042
Supplier:  Eagle Biosciences
Supplier Number:  C2E31-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human CD62E / ELAM-1 / E-Selectin ELISA Assay Kit
  • Size:
    1 kit
  • Cat. No.:
    102994-042
  • Supplier no.:
    C2E31-K01

Specifications

About this item

Human CD62E ELISA detects soluble ELAM-1, (CD62E) in various samples.

  • Dynamic Range: 1-32 ng/ml
  • Sensitivity: 0.5 ng/ml
  • Incubation: 2h10

The Eagle Biosciences Human CD62E ELISA Assay Kit is to be used for the in-vitro quantitative determination of soluble Endothelial Leukocyte Adhesion Molecule ELAM-1, (CD62E) in human serum, plasma, buffered solutions or cell culture medium. The assay will recognize both natural and recombinant human CD62E. The Eagle Biosciences Human CD62E ELISA Assay kit is for Research Use Only.