Etanercept ELISA Assay Kit, Eagle Biosciences
103389-208
:
- Description:Etanercept ELISA Assay Kit
- Size:1 kit
- Cat. No.:103389-208
Etanercept ELISA is for the precise analytical determination of free Etanercept in serum and plasma.
- Dynamic Range: 6 - 200 ng/mL
- Sensitivity: 5 ng/mL
- Incubation: 2 hours
The Eagle Biosciences Etanercept ELISA Assay Kit is an enzyme immunoassay for the precise analytical determination of free Etanercept in serum and plasma. The Etanercept ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.