Order Entry
United States
ContactUsLinkComponent
Colorimetric Nitric Oxide Assay Kit, Eagle Biosciences, Inc.
Colorimetric Nitric Oxide Assay Kit, Eagle Biosciences, Inc.
Catalog # 102994-820
Supplier:  Eagle Biosciences
CAS Number:  
Colorimetric Nitric Oxide Assay Kit, Eagle Biosciences, Inc.
Catalog # 102994-820
Supplier:  Eagle Biosciences
Supplier Number:  NSC31-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Colorimetric Nitric Oxide Assay Kit
  • Size:
    1 kit
  • Cat. No.:
    102994-820
  • Supplier no.:
    NSC31-K01

Specifications

About this item

Nitric Oxide Synthase Colorimetricdetermines Nitric Oxide Synthase in biological fluids.

  • Dynamic Range: 0.5 - 100 µM
  • Sensitivity: 1 pmol/µL or ~1 µM
  • Incubation: 5 minutes

The Nitric Oxide Synthase Colorimetric Assay kit employs immunoaffinity purified Nitrate Reductase (NaR) enabling the measure of total nitric oxide (NO) produced in in vitro experimental systems. Nitric oxide can be spectrophotometrically assayed by measuring the accumulation of its stable degradation products, nitrate and nitrite. The ratio of these two products in biological fluids, tissue culture media, etc. may vary substantially. Hence, for accurate assessment of the total nitric oxide generated, one must monitor both nitrate and nitrite. An excellent solution to this problem is the enzymatic conversion of nitrate to nitrite by the enzyme nitrate reductase, followed by quantitation of nitrite using Griess Reagent. In addition to providing all necessary components in a microtiter format, this kit employs affinity purified nitrate reductase and NADH, thereby circumventing some of the potential problems reported for NO measurement using NADPH dependent nitrate reductases.