Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Primary antibody reactivity:Human
- Target protein:IL-12 (p70)
- Description:Human IL-12p70 ELISA KIT
- Size:1 kit
- Cat. No.:102994-742
- Supplier no.:L7031-K01
Human IL-12p70 EIA detects IL-12p70 concentrations in cell culture supernates,serum, and plasma.
- Dynamic Range: 31.25 - 1000 pg/ml
- Sensitivity: 7 pg/ml
- Incubation: 3.5 hours
The Eagle Biosciences Human Interleukin 12p70 (IL-12p70) ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of human Interleukin 12p70 (IL-12p70) concentrations in cell culture supernates, serum, and plasma. The Eagle Biosciences Human Interleukin 12p70 (IL-12p70) ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.