Order Entry
United States
ContactUsLinkComponent
Human IL-1β ELISA KIT, Eagle Biosciences, Inc.
Human IL-1β ELISA KIT, Eagle Biosciences, Inc.
Catalog # 102994-720
Supplier:  Eagle Biosciences
CAS Number:  
Human IL-1β ELISA KIT, Eagle Biosciences, Inc.
Catalog # 102994-720
Supplier:  Eagle Biosciences
Supplier Number:  L1B31-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-1β
  • Description:
    Human IL-1β ELISA KIT
  • Size:
    1 kit
  • Cat. No.:
    102994-720
  • Supplier no.:
    L1B31-K01

Specifications

About this item

Human Interleukin-1 Beta ELISA detects IL-1 Beta concentrations in biological samples.

  • Dynamic Range: 7.8 - 250 pg/ml
  • Sensitivity: 2 pg/ml
  • Incubation: 3.5 hours

Interleukin-1 (IL-1), originally described as lymphocyte activating factor (LAF) for its effects on thymocytes (1), is a polypeptide cytokine with two molecular forms. The two distinct molecular forms of IL-1 are thought to be derived from two genes (2,3). After transcription, as 31kD precursor polypeptide is cleaved to give rise to mostly cell membrane associated IL-1a and secreted IL-1b. Both have the same molecular weight of 15kD but have different isoelectric points of 5 and 7, respectively.