- Description:H pylori Antigens ELISA Assay Kit (Quantitative)
- Size:1 kit
- Cat. No.:102994-468
- Supplier no.:HPY35-K01
Helicobacter pylori Antigen ELISA determines H
- Dynamic Range: 1.9 - 150 ng/ml
- Sensitivity: 0.165 ng/ml
- Incubation: 2 hours
H. pylori (previously known as Campylobacter pyloridis) is a type of bacteria that infects the stomach and is a common cause of peptic ulcers. H. pylori bacteria can be passed from person to person through direct contact with saliva, vomit or fecal matter. H. pylori can also be spread through contaminated food or water.
pylori antigen in fecal samples.