Order Entry
United States
ContactUsLinkComponent
TBARS Fluorometric Assay, Eagle Biosciences
TBARS Fluorometric Assay, Eagle Biosciences
Catalog # 102995-016
Supplier:  Eagle Biosciences
CAS Number:  
TBARS Fluorometric Assay, Eagle Biosciences
Catalog # 102995-016
Supplier:  Eagle Biosciences
Supplier Number:  TBF39-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    TBARS Fluorometric Assay
  • Size:
    1 kit
  • Cat. No.:
    102995-016
  • Supplier no.:
    TBF39-K01

Specifications

About this item

2-Thiobarbituric Acid Reactive Substances Fluorometric detects TBARS in serum, plasma, or urine.

  • Dynamic Range: 0.5 - 20 µM
  • Sensitivity: 0.5 µM
  • Incubation: 30 minutes

2-ThioBarbituric Acid Reactive Substances (TBARS) are naturally present in biological specimens and include lipid hydroperoxides and aldehydes which increase in concentration as a response to oxidative stress.1,2 TBARS assay values are usually reported in malonaldehyde (malondialdehyde, MDA) equivalents, a compound that results from the decomposition of polyunsaturated fatty acid lipid peroxides. The TBARS assay is a well-recognized, established method for quantifying these lipid peroxides, although it has been criticized for its reactivity towards other compounds other than MDA.3 This TBARS Cuvette Assay Kit offers the researcher a straightforward, reproducible and consistent method for analyzing urine for lipid peroxidation products.