Order Entry
United States
ContactUsLinkComponent
Human IL-22 ELISA KIT, Eagle Biosciences, Inc.
Human IL-22 ELISA KIT, Eagle Biosciences, Inc.
Catalog # 102994-724
Supplier:  Eagle Biosciences
CAS Number:  
Human IL-22 ELISA KIT, Eagle Biosciences, Inc.
Catalog # 102994-724
Supplier:  Eagle Biosciences
Supplier Number:  L2231-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-22
  • Description:
    Human IL-22 ELISA KIT
  • Size:
    1 kit
  • Cat. No.:
    102994-724
  • Supplier no.:
    L2231-K01

Specifications

About this item

Human IL-22 ELISA detects human IL-22 concentrations in cell culture supernates, serum, and plasma.

  • Dynamic Range: 31.25 - 1000 pg/ml
  • Sensitivity: 15 ng/ml
  • Incubation: 3.5 hours

The Eagle Biosciences Human Interleukin 22 (IL-22) ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of human Interleukin 22 (IL-22) concentrations in cell culture supernates, serum, and plasma. The Eagle Biosciences Human Interleukin 22 (IL-22) ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.