Order Entry
United States
ContactUsLinkComponent
Mouse IL-5 ELISA KIT, Eagle Biosciences
Mouse IL-5 ELISA KIT, Eagle Biosciences
Catalog # 102993-504
Supplier:  Eagle Biosciences
CAS Number:  
Mouse IL-5 ELISA KIT, Eagle Biosciences
Catalog # 102993-504
Supplier:  Eagle Biosciences
Supplier Number:  IL511-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IL-5
  • Description:
    Mouse IL-5 ELISA KIT
  • Size:
    1 kit
  • Cat. No.:
    102993-504
  • Supplier no.:
    IL511-K01

Specifications

About this item

Mouse Interleukin 5 (IL-5) ELISA detects IL-5 concentrations in biological samples.

  • Dynamic Range: 15.625 - 500 pg/ml
  • Sensitivity: 7 pg/ml
  • Incubation: 3.5 houea

Mouse interleukin 5 (IL-5), also known as T-cell replacing factor, B-cell growth factor II, eosinophil differentiation factor and eosinophil colony stimulating factor, is a pleiotropic cytokine produced primarily by T cells (1-4). It supports the proliferation and differentiation of mouse, but not human, B cells, and enhances IgM, IgG1, IgA, and IgE secretion. IL-5 chemoattracts and enhances the survival and the effector functions of mature eosinophils and synergizes with various colony stimulating factors to increase eosinophil progenitor production and eosinophil expansion. Because of its diverse effects on eosinophils, IL-5 is strongly implicated in the pathogenesis of asthma and other hypereosinophilic inflammatory conditions (4-6). Mouse IL-5 cDNA encodes a 133 amino acid (aa) residue precursor protein containing a hydrophobic signal peptide that is cleaved to yield a 113 aa residue mature protein. Native mouse IL-5 is a disulfide-linked homodimeric 40-45 kDa glycoprotein with N- and O-linked carbohydrate chains. Mature human IL-5 is approximately 70% identical at the amino acid level to mouse IL-5. Whereas mouse and human IL-5 are equally active on human cell lines, human IL-5 is much less active than mouse IL-5 in mouse cell assays (1-3). IL-5 is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes