AFP ELISA, Eagle Biosciences, Inc.
Catalog # 103389-070
Supplier: Eagle Biosciences
CAS Number:
Specifications
- Target protein:AFP
- Description:AFP ELISA
- Size:1 kit
- Cat. No.:103389-070
Specifications
About this item
AFP ELISA is a colorimetric method to determine Alpha Fetoprotein concentration in serum or plasma.
- Dynamic Range: 5 -200 ng/ml
- Sensitivity: 0.35 ng/ml
- Incubation: 1.5 hours
Alpha Fetoprotein (AFP) is a 68 kDa glycoprotein, which is normally only produced in the fetus during its development. It is a normally produced by the liver and yolk sac of the fetus. AFP levels decrease soon after birth and probably has no function in normal adults. It binds the hormone estradiol to keep it from affecting the fetal brain. Its measurement during pregnancy has been useful to detect certain abnormalities - specifically, if high levels of AFP are found in amniotic fluid, it can indicate a developmental defect in the baby.