Anti Beta 2 Glycoprotein 1 Screen ELISA, Eagle Biosciences
103389-130
:
- Target protein:ANTI BETA2 GLYCOPROTEIN1 IgG
- Description:ANTI BETA 2 GLYCOPROTEIN 1 SCREEN ELISA
- Size:1 kit
- Cat. No.:103389-130
GLP-1 Screen ELISA detect IgG/IgM class antibodies against Beta 2-Glycoprotein 1 in serum or plasma.
- Dynamic Range: 10 - 160 AU/ml
- Sensitivity: 0.47 AU/ml
- Incubation: 2 hours
The antiphospholipid syndrome (APS) is a disorder that presents peculiar symptoms: arterial and venous thrombosis, thrombocytopenia, ulcers of the lower limbs, hemolytic anemia, loss of the fetus during pregnancy and is associated with the presence of antiphospholipid antibodies. Antiphospholipid antibodies represent a large and heterogeneous immunoglobulins group, including anticardiolipin antibodies and lupus anticoagulant. The former are diagnosed by on their reactivity with cardiolipin or other anionic phospholipids in ELISA test, while the latter is detected in phospholipid-dependent coagulation tests (aPTT, KCT, dRVVT).