Order Entry
United States
ContactUsLinkComponent
Potential Antioxidant Capacity Assay, Eagle Biosciences, Inc.
Potential Antioxidant Capacity Assay, Eagle Biosciences, Inc.
Catalog # 102994-838
Supplier:  Eagle Biosciences
CAS Number:  
Potential Antioxidant Capacity Assay, Eagle Biosciences, Inc.
Catalog # 102994-838
Supplier:  Eagle Biosciences
Supplier Number:  PAC39-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Potential Antioxidant Capacity Assay
  • Size:
    1 kit
  • Cat. No.:
    102994-838
  • Supplier no.:
    PAC39-K01

Specifications

About this item

Antioxidant Capacity Potential detects of adduct antioxidant capacity in biological samples.

  • Dynamic Range: 21.9 - 4378 µmol/L (cupric ion reducing power)
  • Sensitivity: 0.063 mM
  • Incubation: 3 minutes

Oxidative stress plays on important role in various diseases and aging. The control of oxidative stress is expected to be useful to prevent diseases and aging. Oxidative stress is caused by the imbalance between reactive oxygen species (ROS) and antioxidant defense system. For accurate assessment of oxidative stress, measurement of ROS, oxidative damage and antioxidant activity may be essential. The Eagle Biosciences Potential Antioxidant Capacity Assay can detect not only hydrophilic antioxidants such as Vitamin C and glutathione, but also can detect hydrophobic antioxidants such as Vitamin E. This Antioxidant Capacity Potential Assay kit is applicable for assessment of total antioxidants of serum, foods and beverage samples.