Order Entry
United States
ContactUsLinkComponent
Nitric Oxide Synthase Non-Enzymatic Assay Kit, Eagle Biosciences
Nitric Oxide Synthase Non-Enzymatic Assay Kit, Eagle Biosciences
Catalog # 102994-816
Supplier:  Eagle Biosciences
CAS Number:  
Nitric Oxide Synthase Non-Enzymatic Assay Kit, Eagle Biosciences
Catalog # 102994-816
Supplier:  Eagle Biosciences
Supplier Number:  NOS39-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Nitric Oxide Synthase Non-Enzymatic Assay Kit
  • Size:
    1 kit
  • Cat. No.:
    102994-816
  • Supplier no.:
    NOS39-K01

Specifications

About this item

Nitric Oxide Synthase Colorimetric Non-Enzymatic detects Nitric Oxide Synthase in biological fluids.

  • Dynamic Range: 0.5 - 100 µM
  • Sensitivity: 1 pmol/µL or ~1 µM
  • Incubation: 5 minutes

Nitric oxide can be spectrophotometrically assayed by measuring the accumulation of its stable degradation products, nitrate and nitrite. The ratio of these two products in biological fluids, tissue culture media, etc. may vary substantially. Hence, for accurate assessment of the total nitric oxide generated, one must monitor both nitrate and nitrite. An excellent solution to this problem involves the conversion of nitrate to nitrite catalyzed by cadmium. This is then followed by the quantitation of nitrite using the griess reaction. This kit is suitable for the quantitative determination of total nitric oxide in samples that have high protein concentrations. The cadmium catalyst proves far more robust when in the presence of harsh deproteinating reagents than the alternative, nitrate reductase.