Order Entry
United States
ContactUsLinkComponent
Mouse Anti Endothelin Receptor A IgG Anti-body ELISA, Eagle Biosciences
Mouse Anti Endothelin Receptor A IgG Anti-body ELISA, Eagle Biosciences
Catalog # 102994-338
Supplier:  Eagle Biosciences
CAS Number:  
Mouse Anti Endothelin Receptor A IgG Anti-body ELISA, Eagle Biosciences
Catalog # 102994-338
Supplier:  Eagle Biosciences
Supplier Number:  ERG11-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugate ELISA:
    HRP
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse Anti Endothelin Receptor A IgG Antibody ELISA
  • Size:
    1 kit
  • Cat. No.:
    102994-338
  • Supplier no.:
    ERG11-K01

Specifications

About this item

Mouse Anti Endo Receptor A IgG Antibody ELISA detect Abs against Endothelin receptor subtype A.

  • Dynamic Range: 1.25 -40 U/ml
  • Sensitivity: Cut-off
  • Incubation: 3.5 hours

Endothelins (ET) are 21-amino acid vasoconstricting peptides produced primarily in the endothelium having a key role in vascular homeostasis. It mediates the effects through G-Protein-coupled receptors, the Endothelin receptors. There are two key receptor types, ETA and ETB. ETA receptors are found in the smooth muscle tissue of blood vessels where they increase vasoconstriction by ET-1.

The Eagle Biosciences Mouse Anti Endothelin Receptor A IgG Antibody is designed for the determination of antibodies (IgG) against the Endothelin receptor subtype A in serum and plasma.