Order Entry
United States
ContactUsLinkComponent
CXCL 12/SDF-1 Alpha ELISA, Eagle Biosciences
CXCL 12/SDF-1 Alpha ELISA, Eagle Biosciences
Catalog # 102994-140
Supplier:  Eagle Biosciences
CAS Number:  
CXCL 12/SDF-1 Alpha ELISA, Eagle Biosciences
Catalog # 102994-140
Supplier:  Eagle Biosciences
Supplier Number:  CXC31-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    CXCL 12/SDF-1 Alpha ELISA
  • Size:
    1 kit
  • Cat. No.:
    102994-140
  • Supplier no.:
    CXC31-K01

Specifications

About this item

CXCL 12/SDF-1 Alpha ELISA is used to determine CXCL 12 / SDF-1 Alpha in serum and plasma.

  • Dynamic Range: 0.625 - 40 ng/ml
  • Sensitivity: 0.31 ng/ml
  • Incubation: 2.5 hours

SDF-1 Alpha (CXCL12) is a CXC chemokine [1]. The small basic protein (ca. 8 kDa) has no glycosylation site in its sequence [2]. The homology properties of SDF-1 Alpha are unique among the chemokines. It has a high degree of conservation within the mammalians, with 99 % identity between the human and the murine SDF-1 Alpha sequences. But it shows a weak homology to the other CXC chemokines and the SDF-1 gene is on chromosome 10, whereas the other chemokine genes are on chromosome 4 and 17 [3].