Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Target protein:Trail Receptor 1/DR4/CD261
- Description:Human Trail Receptor 1/DR4/CD261 ELISA Assay Kit
- Size:1 kit
- Cat. No.:102994-066
- Supplier no.:C6131-K01
Human TRAIL R1/DR4 ELISA detects TRAIL R1/DR4 in biological fluids.
- Dynamic Range: 31.25-1000 pg/ml
- Sensitivity: 8 pg/ml
- Incubation: 2h30
The Eagle Biosciences Human TRAIL R1/DR4 ELISA is a solid phase sandwich ELISA for the in-vitro qualitative and quantitative determination of TRAIL R1/DR4 in cell culture supernatants, buffered solutions or human serum, plasma, or other body fluids. This assay will recognize both natural and recombinant human TRAIL R1/DR4. The Eagle Biosciences Human TRAIL R1/DR4 ELISA Assay kit is for Research Use Only.