Bevacizumab ELISA Assay Kit, Eagle Biosciences, Inc.
103389-212
:
- Description:Bevacizumab ELISA Assay Kit
- Size:1 kit
- Cat. No.:103389-212
Bevacizumab ELISA analyzes biologically active form of free bevacizumab in serum and plasma samples.
- Dynamic Range: 6 -200 ng/mL
- Sensitivity: 2 ng/mL
- Incubation: 2 hours
The Eagle Biosciences Bevacizumab (Avastin®) ELISA Assay Kit is an enzyme immunoassay for the precise analytical determination of free Bevacizumab in serum and plasma samples. The Bevacizumab (Avastin®) ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.