Order Entry
United States
ContactUsLinkComponent
Human Trail Receptor 4/DcR2/CD264 ELISA Assay Kit, Eagle Biosciences
Human Trail Receptor 4/DcR2/CD264 ELISA Assay Kit, Eagle Biosciences
Catalog # 102994-072
Supplier:  Eagle Biosciences
CAS Number:  
Human Trail Receptor 4/DcR2/CD264 ELISA Assay Kit, Eagle Biosciences
Catalog # 102994-072
Supplier:  Eagle Biosciences
Supplier Number:  C6431-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Target protein:
    Trail Receptor 4/DcR2/CD264
  • Description:
    Human Trail Receptor 4/DcR2/CD264 ELISA Assay Kit
  • Size:
    1 kit
  • Cat. No.:
    102994-072
  • Supplier no.:
    C6431-K01

Specifications

About this item

Human TRAIL R4/DcR2 ELISA detects TRAIL R4/DcR2 in biological samples

  • Dynamic Range: 312.5-10000 pg/ml
  • Sensitivity: 64 pg/ml
  • Incubation: 3h40

The Eagle Biosciences Human TRAIL R4/DcR2 ELISA Assay is a solid phase sandwich ELISA for the in-vitro qualitative and quantitative determination of TRAIL R4/DcR2 in cell culture supernatants, buffered solutions or human serum, plasma, or other body fluids. This assay will recognize both natural and recombinant human TRAIL R4/DcR2. The Eagle Biosciences Human TRAIL R4/DCR2 ELISA Assay kit is for Research Use Only.