Order Entry
United States
ContactUsLinkComponent
 
Anti-CaSR IgG ELISA _ DO NEW PROTOCOL, Eagle Biosciences, Inc.
Catalog # 102994-130
Supplier:  Eagle Biosciences
CAS Number:  
Anti-CaSR IgG ELISA _ DO NEW PROTOCOL, Eagle Biosciences, Inc.
Catalog # 102994-130
Supplier:  Eagle Biosciences
Supplier Number:  CSR31-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Anti-CaSR IgG ELISA _ DO NEW PROTOCOL
  • Size:
    1 kit
  • Cat. No.:
    102994-130
  • Supplier no.:
    CSR31-K01

Specifications

About this item

Human Anti-CaSR IgG EIA detects human anti-CaSR IgG autoAb levels in serum, plasma, tissue extract.

  • Dynamic Range: 10 - 270 U/ml
  • Sensitivity: 2.5 U/ml
  • Incubation: 2.0 hours

The Eagle Biosciences Human Anti-CaSR ELISA Assay Kit (enzyme-linked immunoassay kit) kit is produced for the quantitative determination of human anti-CaSR (calcium sensing receptor) autoantibody levels in serum, plasma, tissue extract or other liquid samples.