Order Entry
United States
ContactUsLinkComponent
Human CD86 ELISA Assay Kit, Eagle Biosciences
Human CD86 ELISA Assay Kit, Eagle Biosciences
Catalog # 102994-076
Supplier:  Eagle Biosciences
CAS Number:  
Human CD86 ELISA Assay Kit, Eagle Biosciences
Catalog # 102994-076
Supplier:  Eagle Biosciences
Supplier Number:  C8631-K01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human CD86 ELISA Assay Kit
  • Size:
    1 kit
  • Cat. No.:
    102994-076
  • Supplier no.:
    C8631-K01

Specifications

About this item

Human CD86 ELISA is to be used for in-vitro quantitative determination CD86 in biological samples.

  • Dynamic Range: 1.187 U/ml - 38 U/ml
  • Sensitivity: 0.6 U/ml
  • Incubation: 2.75 hours

The Eagle Biosciences Human CD86 ELISA is to be used for the in-vitro quantitative determination CD86 in human serum, plasma, buffered solutions or cell culture medium. The assay will recognize both natural and recombinant human CD86. The Eagle Biosciences Human CD86 ELISA Assay kit is for Research Use Only.