Order Entry
United States
ContactUsLinkComponent
Hexanoyl-Lys adduct (HEL) ELISA , Eagle Biosciences, Inc.
Hexanoyl-Lys adduct (HEL) ELISA , Eagle Biosciences, Inc.
Catalog # 102994-458
Supplier:  Eagle Biosciences
CAS Number:  
Hexanoyl-Lys adduct (HEL) ELISA , Eagle Biosciences, Inc.
Catalog # 102994-458
Supplier:  Eagle Biosciences
Supplier Number:  HEL39-K01
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay Type:
    Competitive
  • Description:
    Hexanoyl-Lys adduct (HEL) ELISA
  • Size:
    1 kit
  • Cat. No.:
    102994-458
  • Supplier no.:
    HEL39-K01

Specifications

About this item

The Hexanoyl-Lys adduct (HEL) EIA is intended to determine HEL adduct in biological samples.

  • Dynamic Range: 2.6 - 624 nmol/L
  • Sensitivity: 2.6 nmol/L
  • Incubation: Overnight

Hexanoyl-Lys adduct (HEL) is formed by the reaction of linoleic acid hydroperoxide and Lysine, and a biomarker for oxidative stress. The Hexanoyl­Lysine adduct ELISA assay kit is a competitive enzyme-linked immunosorbent assay for quantitative measurement of hexanoyl-Lys adduct. This kit is based on the monoclonal antibody clone 5H4, which is specific for HEL. Suitable for urine, serum and other biological samples.