The Eagle Biosciences 14, 15-DHET ELISA Assay kit is intended for the quantitative determination of 14, 15-DHET in biological samples by enzyme linked immunoassay (ELISA)
- Measure 14,15-DHET in biological samples
- Intended for the quantitative determination of 14,15-DHET
- Competitive ELISA
The Eagle Biosciences 14, 15-DHET ELISA Assay kit is intended for the quantitative determination of 14, 15-DHET in biological samples by enzyme linked immunoassay (ELISA). The Eagle Biosciences 14, 15 DHET ELISA Assay kit is for research use only and not to be used in diagnostic procedures.
The Eagle Biosciences 14, 15 DHET ELISA Assay kit is for research use only and not to be used in diagnostic procedures.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Assay Type:Competitive
- Target protein:14 15-DHET