ContactUsLinkComponent
pBK-CMV Phagemid Vector, Agilent Technologies
  99900-386
 :  Avantor
 :  
undefined
pBK-CMV Phagemid Vector, Agilent Technologies
  99900-386
 :  Avantor
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    pBK-CMV Phagemid Vector
  • Size:
    20 μg
  • Environmentally Preferable:
  • Cat. No.:
    99900-386
  • Supplier no.:
    212209
  • Vector details:
    Allows both prokaryotic and eukaryotic expression.
pBK-CMV vector used to express proteins in both E. coli and mammalian model systems.
  • Constitutive expression in eukaryotic cells
  • Inducible protein expression in prokaryotes
  • Rescued from ZAP Express lambda vector
In mammalian cells, the expression is driven by the CMV promoter and is therefore constitutively expressed. In E. coli, the lac promoter drives high-level expression in the presence of the inducer IPTG, and allows minimal expression in the absence of inducer.
 : Kit contains 20 μg pBK-CMV phagemid vector, 500 μl XL1-Blue MRF, and 1 ml R408 Interference-Resistance helper phage.
 : For research use only. Not for use in diagnostic procedures.