Order Entry
United States
ContactUsLinkComponent
 
Oakton pHTestr 10 Waterproof BNC Pocket pH Tester, Cole-Parmer
  11026-016
undefined
Oakton pHTestr 10 Waterproof BNC Pocket pH Tester, Cole-Parmer
  11026-016
 :  35634-14

 

  • Description:
    Oakton pHTestr 10 Waterproof BNC Pocket pH Tester
  • pH Range:
    0 (min.) – 14 (max.)
  • Cat. No.:
    11026-016
  • For use with:
    Oakton pH electrodes with BNC connector
  • Supplier no.:
    WD-35634-14

 

 

Customizable to your application – use with any pH electrode with a BNC connector.

  • Versatility of a handheld meter at a pocket price
  • IP67-rated protects meter from water, dust, and shock while offering chemical resistance
  • Meter floats, accidental drops into water will not result in lost or damaged meters
 : Includes protective plastic storage case, lanyard, and batteries.