HardyDisks™Cephalothin, CF-30, AST, Hardy Diagnostics
Catalog # 76169-768
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:Cephalothin, CF-30
- Size:1 x 50 Disks
- Cat. No.:76169-768
Specifications
About this item
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing. The HardyDisk™ AST cartridges are designed to be fully compatible with the BBL single or multiple disk dispensers.